TruCLEAN® Mops and Mop Covers, Perfex
Supplier: Perfex
22-33 22-34 22-24 22-36 22-34F 22-35
22940-020CS 601.32 CAD
22940-020 22940-023 22940-304 89096-004 89096-030 22940-191
TruCLEAN® Mops and Mop Covers, Perfex
Mops
Mop heads are ideal for use in controlled environments and for general housekeeping applications
Quick-release frame-locking mechanism allows easy mop head replacement. Mop frames are made of high-grade ANSI stainless steel. The fixed mop frame eliminates swivel movement for hassle-free cleaning of ceilings and walls. Swivel mop frame features a lockable swivel for situations when mopping in an “S” pattern is beneficial.
All components are compatible with gamma, ETO, and autoclave sterilization.
Yarn mop head features looped-end construction and thoroughly cleans a 508cm2 (80 sq. in.More Product Information