BRAND BRANDplates® lipoGrade™ Immunoassay Microplates, Nonsterile
Supplier: Brandtech
781840 781842
89093-656PK 478.59 CAD
89093-656 89093-660 89093-670 89093-674 89093-764
BRAND BRANDplates® lipoGrade™ Immunoassay Microplates, Nonsterile
Microplates Immunology Microplates and Strips
BRANDplates® lipoGrade™ plates are nonsterile plates treated for immobilization of predominantly hydrophobic proteins for solid phase assays
  • Binds hydrophobic proteins
  • Maintains hydrophilic proteins in solution
  • ANSI-SBS formatted
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. The 96-Well plates have a blue embossed alphanumeric code for quick and easy well identification.
Convesely, they are also excellently suited for homogeneous assays ofMore Product Information