PCR Mini Cooler, with Transparent Lid, BrandTech®

Supplier: Brandtech

781260
89093-070PK 484.7 CAD
89093-070
PCR Mini Cooler, with Transparent Lid, BrandTech®
Boxes Cooling/Freezing Boxes and Bags
The mini PCR cooler keeps samples at 4 ºC for approximately 3 hours.

  • Maintains 4 ˚C for master mix stability
  • Color indicates temperature
  • Holds 0.2 ml tubes, strips or plates
  • For use with PCR tubes 0.2 mL single, 8-strips, 12-strips and 96-well PCR plates

The insulating gel changes from violet to pink at 7 ºC. Suitable for 0.2 ml single tubes, 8-strips, and 12-strips, as well as 96-well PCR plates.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR