PYREX® Drying Tube with [ST] Joint, Corning
Supplier: Corning
7800-24
89091-578CS 3348.76 CAD
89091-578
PYREX® Drying Tube with [ST] Joint, Corning
Tubes Drying Tubes
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These inverted drying tubes have an inner 24/40 Standard Taper joint at one end only and a single bulb. The chamber is approximately 110 mm long including the bulb (30 mm Ext.Ø) and will take a No. 2 rubber stopper.