PYREX® 9 Depression Spot Plate, Corning
Supplier: Corning
7220-85
89090-482CS 1577.01 CAD
89090-482
PYREX® 9 Depression Spot Plate, Corning
Staining Tiles
These pressed PYREX® glass plates have 9 concave depressions which is 7 mm deep and has 22 mm outer diameter.
The plate is made of borosilicate glass which has size 85×100 mm.