PYREX® VISTA™ Volumetric Flasks with Snap Caps, Class B, Corning
Supplier: Corning
70581-10 70581-1L 70581-100 70581-2L 70581-250 70581-25 70581-500 70581-50
89090-118CS 522.57 CAD
89090-118 89090-300 89090-306 89090-332 89090-344 89090-356 89090-382 89090-394
PYREX® VISTA™ Volumetric Flasks with Snap Caps, Class B, Corning
Flasks Volumetric Flasks
Made of PYREX® glass, with PE snap-cap.
  • Heavy duty necks
  • Graduation line is sharp and permanent
  • Inscriptions easy to read
Designed for use by customers that do not require the precision accuracy of our Class A capacity flasks. The shape, body and wall are the same as regular flasks but display the large blue marking spot and designation 'PYREX VISTA' unique to the PYREX VISTA line. Capacity tolerances are twice those specified for Class A volumetric ware.
Heavy-duty necks are tooled for HDPE snap-caps, which are included with all sizes. FlatMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Monterrey