Corning® Digital Magnetic Hotplate Stirrers, 230 V
Supplier: Corning
6796-620D 6798-620D 6796-420D 6798-420D
89090-200CS 1993.47 CAD
89090-200 89090-202 89090-218 89090-220
Corning® Digital Magnetic Hotplate Stirrers, 230 V
Stirrers Magnetic Hotplate Stirrers
Consistent temperature for your classroom.
  • 230 V model
  • Closed-loop stirring control monitors and regulates the stirring speed for aqueous, viscous, or semi-solid solutions
  • Microprocessor maintains consistent and repeatable temperature settings
  • For digital displays, LED temperature display is adjustable in increments of 5 °C and flashes until set temperature is reached
  • Built-in holder accommodates optional 102 mm stainless steel support rods
  • Stronger, larger magnet improves magnetic stirring bar coupling combined with automatic ramp up speed adjustment
More Product Information