HTS Transwell® 96-Well Permeable Support Systems and Plates, Corning
Supplier: Corning
3391 3380 3392 3385 3386 3388 3387 3374 3381 7369 3384
89089-924CS 2550.59 CAD
89089-924 89089-926 89089-928 89089-930 89089-932 89089-934 89089-936 89089-938 89089-922 10185-052 89089-941
HTS Transwell® 96-Well Permeable Support Systems and Plates, Corning
Microplates Cell Culture Plates
Permeable supports for HTS (high throughput screening) applications are excellent for ADMET screening, drug transport, and other assays.
The HTS Transwell® 96-well permeable support features 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit. Available with polyester or polycarbonate membranes in a variety of pore sizes. The 0.143cm² well membrane area provides significantly more surface area than comparable systems. Large apical and basolateral access ports allow efficient media sampling and facilitateMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk