Corning® Stripette® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged, Corning
Supplier: Corning
4010 4011 4012 4020 4021 4050 4100 4250 4500 4501 4485 4486 4487 4489 4490 7000 7015 7041 7016 7042 7017 7045 4491 4051 4101 4251 4484 4488 4492
29442-406CS 718.13 CAD
29442-406 29442-408 29442-410 29442-416 29442-418 29442-420 29442-428 29442-434 29442-438 29442-440 29443-041 29443-043 29443-045 29443-049 29443-051 89089-882 89089-884 89089-886 89089-888 89089-890 89089-892 89089-894 89091-046 29442-422 29442-430 29442-436 29442-886 29443-047 89091-048
Corning® Stripette® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged, Corning
Pipettes Serological Pipettes
PS, transparent, graduated, sterile.
  • Calibrated to deliver (TD, Ex)
  • High performance anti-drip tips, colour-coded magnifier stripe and negative graduations
  • Certified RNase-/DNase-free and non pyrogenic
* Wide orifice for viscous materials.
Paper/plastic wrap allows easy opening by either pop-through or peel-apart techniques. Ideal for sterile tissue culture applications. Wrap reduces static cling.
Exclusive anti-drip tip assures accurate delivery (25, 50, and 100 ml sizes). Color-coded magnifier strips make volume reading easier. BidirectionalMore Product Information