Corning® Solid Black and White Polystyrene Microplates, 384 Well
Supplier: Corning
3573 3576 3574 3570 3820 3822 3824 3826 3570BC 3825 3821 3821BC 3824BC 4510 4511 4512 4513 4514 3575BC 3574BC
89089-788CS 735.94 CAD
89089-788 89089-794 89089-796 89089-800 89089-856 89089-860 89089-868 89089-870 89176-442 89185-546 89089-858 89203-192 89203-194 89511-310 89511-312 89511-314 89511-316 89511-318 CA89089-786 CA89089-798
Corning® Solid Black and White Polystyrene Microplates, 384 Well
Microplates HTS Microplates
PS. Designed to reduce well-to-well crosstalk during fluorescent and luminescent assays.
  • Square well top for easier robotic pipetting
  • Round bottom for better Z factor and minimized trapped air
  • Conical well molded in the shape of a light cone for efficiency
  • Black walls reduce well-to-well crosstalk and autofluorescence during fluorescent assays
High binding surface is a general purpose surface that binds medium (>10kD) and large biomolecules possessing ionic groups and/or hydrophobic regions. Binding capacity is approximately 400 to 500ng IgG/cm.