Corning® Polycarbonate Erlenmeyer Flasks with Screw Caps
Supplier: Corning
431252 431256 431253 431255 430421 430183 430422 431144 431145 431143 431146 431147 431404 431405 431402 431403 431406 431407 431408 431401 431684 431685
17777-100CS 568.74 CAD
17777-100 17777-102 17777-104 17777-106 29152-146 29152-168 29152-180 29445-056 29445-058 29445-060 29445-062 29445-064 89089-716 89089-718 89089-750 89089-752 89089-758 89089-760 89089-810 89089-812 10098-584 10098-586
Corning® Polycarbonate Erlenmeyer Flasks with Screw Caps
Flasks Erlenmeyer Flasks
PC, optically clear.
  • Baffled or plain bottom options in all sizes (125 ml to 3 L)
  • Moulded-in graduations for accuracy
  • Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
  • Individually packaged and radiation sterilised for ease of use
  • Certified non-pyrogenic
Corning® baffled and plain Erlenmeyer flasks are ideal for shaker culture applications and storage.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleteriousMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Oneonta RECs, ISO 14001 - Oneonta