Finntip Flex Pipette Tips, Thermo Scientific
Supplier: Thermo Fisher Scientific
94060813 94060713 94060717 94060720 94060820 94060970 94060310 94060313 94060320 94060120 94060100 94060103 94060117 94060817 94060810 94060317
11388-564CS 459.98 CAD
11388-564 89047-118 89047-120 89047-122 89047-124 89047-126 89049-822 89087-470 89087-472 89087-474 89087-478 89087-484 89087-486 89087-488 89087-490 89087-492 89087-494 89087-496 11388-562 89087-476
Finntip Flex Pipette Tips, Thermo Scientific
Pipette Tips
The latest breakthrough in pipette tip technology, the Finntip Flex system offers flexible, soft and sophisticated high-end tips for discerning users.
Thermo Scientific Finntip Flex pipette Tips ensure a better seal with the pipetter module, for higher accuracy and precision. The Finntip Flex system consists of a new generation of easy-to-use 10, 200, 300, 1000, 1200 µl and 10 ml tips.
The soft tip reduces the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI). It also ensures a better seal with the pipetter module,More Product Information