Clamp, Dual Adjustment, Ace Glass Incorporated
Supplier: Ace Glass
11073-20 11073-27
89083-242EA 118.55 CAD
89083-242 89083-244
Clamp, Dual Adjustment, Ace Glass Incorporated
Clamps General Purpose Clamps
This clamp has a dual adjustment feature
It is designed so that the vinylized jaws open at a gradual angle, to their extreme position, without binding. The screws, wing-nuts, and pivot pins are nickel-plated; springs are stainless steel. The extension arm is a seamless tube, nickel-plated.