Trubore® Bearing, Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
8066-320 8066-324 8066-328 8066-333 8066-332 8066-08 8066-220 8066-224 8066-228 8066-233 8066-232 8066-03
89082-928EA 941.02 CAD
89082-928 89082-930 89082-932 89082-934 89082-936 89082-948 89084-796 89084-798 89084-800 89084-802 89084-804 89084-924
Trubore® Bearing, Ace-Thred, Ace Glass Incorporated
Stirrers Stirring Tools
This combination, all-in-one, stirring shaft bearing and debris trap is designed to prevent particles generated by bearing wear from entering the reaction vessel
  • A lubricant-free PTFE bearing with compression saddle O-Ring creates a mechanical seal against a polished glass shaft
  • The debris trap section consists of a PTFE washer and particle well
  • Fabricated from borosilicate glass
A #7 Ace-Thred on the side of the glass body allows easy access for clean out, or it may be used as an evacuation/gas purge port.
Delivery information: Available as aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable