Hose Connection Adapter, Bent, Ace Glass Incorporated
Supplier: Ace Glass
5205-05 5205-10 5205-25 5205-35 5205-15 9088-09 5205-110 5205-112 5205-114 9088-07 5205-120 5205-124 5205-16
89051-402EA 81.42 CAD
89051-402 89051-420 89051-430 89051-434 89051-510 89064-866 89082-630 89082-632 89082-634 89064-864 10098-736 10098-738 10767-890
Hose Connection Adapter, Bent, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These Hose Connection Adapters are available with either a standard taper inner or a spherical ball bottom joint.
  • Inner [ST] or spherical ball bottom joint
  • 90° angle top hose connection
  • Fabricated from borosilicate glass
At the top of the adapter is a bent 90° hose connection. Fabricated from borosilicate glass. Also available with an integral Porosity A (145–174 micron) fritted disc.
Order Now
SPECIFICATIONS
Environmentally Preferable