OneTouch™ Pipette Tips, Sorenson BioScience
Supplier: Sorenson Bioscience
10300 10310 10330 10340 10360 10370 10390 10410
89082-334CS 855.78 CAD
89082-334 89082-336 89082-340 89082-342 89082-346 89082-348 89082-352 89082-356
OneTouch™ Pipette Tips, Sorenson BioScience
Pipette Tips
Each year 4000000 lbs. of plastic from pipette tip racks are deposited in landfills. e.dek offers a convenient recycling solution that eliminates plastic rack waste. Using a dual shot technology, combining TPE with polypropylene, we are pleased to announce a revolution in liquid handling. OneTouch tips retain all the accuracy and delivery performance that comes with a full polypropylene tip.
  • 100% recyclable
  • Biodegradable
  • Leak proof seal
  • Minimal force required to eject tip
  • Graduated and extended volume
  • Certified RNase/DNase-free
The TPE soft collarMore Product Information