VWR® Inkjet / VWR® Inkjet Plus Microscope Slides, Premium
Supplier: VWR International
89078-508 89078-501 89078-519 89078-523 89078-525 89078-527 89078-531 89078-533 89078-535
89078-508CS 1481.7 CAD
89078-508 89078-501 89078-519 89078-523 89078-525 89078-527 89078-531 89078-533 89078-535
VWR® Inkjet / VWR® Inkjet Plus Microscope Slides, Premium
Slides Microscope Slides
Slides have a frosted tab which is designed to be directly printed upon by Leica® Microsystems IP S ink jet printer.
  • Available in various colors
  • Frosted edges
Ideal for histology, cytology, hematology, and microbiology applications. Frosted tabs are available in various colors. Inkjet Plus slides include a plus charge symbol.