Stirrer Magnets, Disposable, Ace Glass Incorporated
Supplier: Ace Glass
13659-14 13659-18 13659-22 13659-27
89077-982PK 590.43 CAD
89077-982 89077-984 89077-986 89077-988
Stirrer Magnets, Disposable, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
Stirrer magnets are designed for single use applications where efficiency and cross contamination is a concern
Manufactured from Alnico V, these magnets have a PTFE coating that is inert for high purity contact.