Filter Regulator and Lubricator Multple Unit, Ace Glass
Supplier: Ace Glass
13372-45
89077-666EA 781.95 CAD
89077-666
Filter Regulator and Lubricator Multple Unit, Ace Glass
Gas Regulators
Space-saving, multiple unit, recommended for use with certain ACE air motors
Pressurized air flows through louvered deflector in swirling pattern, with liquids and dirt falling into lower baffle where they are prevented from reentering the air stream. Element removes impurities down to 40 microns. Clean air then passes through precision needle valve feed mist lubricator that can be filled under pressure. 150psi maximum pressure range is adjustable through spring action of T-handle. Maximum operating temperature is 125°F. Manual drain. Shatter-proof polycarbonate bowls not recommended forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable