Air Stirrer, Light Duty, Ace Glass
Supplier: Ace Glass
13365-05
89077-660EA 1376.03 CAD
89077-660
Air Stirrer, Light Duty, Ace Glass
Stirrers Overhead Stirrers
The small, compact, and quiet air motor is ideal for stirring all types of solvents, lacquers, paints, oils, synthetics, and fine and heavy chemicals
The device requires an air supply of only 30 to 100PSI. Variable speed ranges from 200rpm to 10,000rpm simply by turning the air supply line valve. For bath sizes to 20L, this smooth running unit has little or no service cost. Unit adjusts itself to compensate for wear. The shaft and propeller are made of stainless steel to resist most acids and chemicals, and for easier cleaning. The device will start in most stalled positions with low airMore Product Information