Tripod Support Stand for 100 and 150 mm Columns, Ace Glass Incorporated
Supplier: Ace Glass
12099-20
89076-384EA 597.46 CAD
89076-384
Tripod Support Stand for 100 and 150 mm Columns, Ace Glass Incorporated
Supports Flask Supports
Tripod support for bottom of Ion-exchange columns
Metal support has hole large enough for end fitting to pass through so that shoulder of column rests on plate. Column should also be stabilized with a clamp. For use only with 100 and 150 mm diameter columns.