Instatherm® Low-Form Oil Bath, Ace Glass Incorporated
Supplier: Ace Glass
9601-08 9601-12 9601-14 9601-16 9601-18 9601-38 9601-42 9601-44 9601-46 9601-48
89075-322EA 1631.69 CAD
89075-322 89075-324 89075-326 89075-328 89075-330 89075-348 89075-350 89075-352 89075-354 89075-356
Instatherm® Low-Form Oil Bath, Ace Glass Incorporated
Baths Oil Baths
Low form, borosilicate glass open vessels coated with Ace Instatherm®
Caution: Never run Instatherm® baths dry or breakage can occur, especially at higher temperatures.
Designed to operate up to 250°C, heating response time is very rapid, (5° per minute), with very low thermal lag. The bottom is uncoated so the bath can be put on any popular magnetic stirrer for stirring while heating. Baths are designed to run with oil or a heating media. Capacity listed is without a vessel. Three-prong connector plug is designed to work with Ace temperature controllers where the output voltage can beMore Product Information