PIG® Oil-Only Absorbent Mat Roll, New Pig
Supplier: New Pig
MAT401 MAT419 MAT424 MAT425 MAT461 MAT462 MAT441
56620-066EA 290.51 CAD
56620-066 89073-844 89073-848 89073-850 89073-856 89073-858 89205-906
PIG® Oil-Only Absorbent Mat Roll, New Pig
Spill Control Materials
Eight layers of 100% polypropylene are thermally bonded to give PIG Mat excellent strength; won't rip, tear or fray even when saturated
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job
  • Rolls are ideal for covering large areas and soaking up leaks and overspray
  • Easy-tear perforations let you take only what you need, so you use less mat and save money
Heavyweight construction is highly durable andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable