VWR® Reclosable Parts Bags
Supplier: VWR International
89071-890 89071-892 89071-896
89071-890CS 184.2 CAD
89071-890 89071-892 89071-896
VWR® Reclosable Parts Bags
Bags Storage Bags
These bags have been chosen for their ability to rival any reclosable bag at economical prices.
  • Full gauge
  • Exceptional clarity and quality
  • Hanging hole
  • Prepared writing area
These reclosable bags are full gauge and offer exceptional clarity and quality. They feature a large white writing area that accepts either marker or pen and a single hang hole above the zipper closure for a secure hang. These tough bags employ a reinforced side-welded zipper to lock in single smooth motion. Made of 100% virgin Low Density Polyethylene (LDPE) resin.