Sanitary Fitting to Hose Barb Fitting Adapter, Colder Products Company
Supplier: Colder Products
SQCC222424M SQCX221212M
89066-556EA 1320.33 CAD
89066-556 89066-558 89066-560 89066-562
Sanitary Fitting to Hose Barb Fitting Adapter, Colder Products Company
Fittings and Connectors
Stainless steel adapter engineered to ease the transition from stainless steel process pipe and equipment to single use disposable plastic.
Materials of construction include 316L stainless steel and platinum cured, USP class 6 silicone oring seals.
Certifications: USP Class VI, BSE/TSE free materials (ADCF)
Ordering information: Accessories not included
Order Now
SPECIFICATIONS
Environmentally Preferable