Automatic Pressure Release Valve, Adjustable, Ace Glass Incorporated
Supplier: Ace Glass
8769-10 8769-110 8769-20
89064-032EA 354.67 CAD
89064-032 89064-034 89064-036
Automatic Pressure Release Valve, Adjustable, Ace Glass Incorporated
Valves Vacuum Valves
Automatic, adjustable pressure release valve for safer operation of pilot plant or other assemblies where the pressure is of great concern
The tension on the stainless steel spring determines the amount of pressure needed to force the lower piece open. With the spring supplied, the range is 15–50 psi. Available with or without warning whistle. Plunger made of PTFE with FETFE o-rings.