Bubbler, Mineral Oil, Adjustable, with Ace-Threds, Ace Glass Incorporated
Supplier: Ace Glass
8762-08 8762-20 8762-35 8762-05
89063-996EA 79.75 CAD
89063-996 89064-000 89064-002 89063-992
Bubbler, Mineral Oil, Adjustable, with Ace-Threds, Ace Glass Incorporated
Bubblers
This mineral oil bubbler is equipped with a completely adjustable 7mm inner tube
Volume of the bubbler is approximately 40mL. Tapered bottom of vessel allows for use with small volumes. A #7 Ace-Thred nylon bushing (89051-122) is supplied, along with a FETFE O-ring. An optional inner tube with check valve (89064-002) is also available.
The bubbler vessel and inner tube are manufactured from borosilicate glass.
The tube has a pulled tip, and is secured to the vessel via a #7 Ace-Thred bushing with FETFE O-ring (89051-122). The inlet and outlet are also #7 Ace-Thred joints, suppliedMore Product Information