Tissue Homogenizer, Ace Glass Incorporated
Supplier: Ace Glass
8325-04 8325-06 8325-08 8325-16 8325-18 8325-20 8325-22 8325-24 8325-26 8325-28 8325-30 8325-32
89062-994EA 206.07 CAD
89062-994 89062-996 89062-998 89063-000 89063-002 89063-004 89063-006 89063-008 89063-010 89063-012 89063-014 89063-016
Tissue Homogenizer, Ace Glass Incorporated
Homogenizers Tissue Grinders
This highly efficient, all glass, tapered tissue grinder/homogenizer combines two grinding surfaces on a single pestle and tube
The conical area is for initial grinding, and the cylindrical area is for final homogenization. Interchangeably ground to 0.1–0.15 mm clearance. With pouring lip. The 10 mm rod on the three larger size pestles may be used with the 89062-242 "Flex-Grip" chuck. The 6 mm rod on the small size is for use with the 89062-238 chuck. Capacities represent volumes with pestle inserted. Fabricated from borosilicate glass.
Ordering information: Tube and pestle may beMore Product Information