Stirring Shaft Coupling, Ace Glass Incorporated
Supplier: Ace Glass
8126-05 8126-08 8126-10 8126-19 8126-22 8126-24 8126-28 8126-01 8126-02 8126-03
89062-282EA 376.77 CAD
89062-282 89062-284 89062-286 89062-288 10767-332 10767-334 10767-942 76318-576 76318-578 76318-580
Stirring Shaft Coupling, Ace Glass Incorporated
Stirrers Stirring Tools
Universal swivel coupling for connection to a metal chuck
  • Stainless steel stir shaft couplings connect glass or metal stir shafts to the adjustable chuck on an overhead stir motor
  • Compression connection is secured to various O.D. shafts via an Allen screw coupling
Pass-thru assemblies contain coupling, ⅜" union, and ⅜×12" extension shaft.
The plastic compression connection, secured via an Allen screw, attaches to various O.D. glass stirring shafts. When used with pass-through assemblies, the coupling allows for easy height adjustment flexibilityMore Product Information