Stirrer Blades, PTFE, for Button-Type Shafts, Ace Glass Incorporated
Supplier: Ace Glass
8082-02 8082-04 8082-06 8082-08 8082-100
89062-054EA 30.02 CAD
89062-054 89062-056 89062-058 89062-060 89062-062
Stirrer Blades, PTFE, for Button-Type Shafts, Ace Glass Incorporated
Stirrers Stirring Blades
Oval-shaped PTFE stirrer blades are designed for use with 10mm O.D
button-type stirring shafts.
Ordering information: Item 89062-062 is supplied as a pack of 6 blades, with two each of blades 89062-054 and 89062-056, and one each of blades 89062-058 and 89062-060.
Order Now
SPECIFICATIONS
Environmentally Preferable