Bearing, Trubore®, PTFE, 19 mm, Ace Glass Incorporated
Supplier: Ace Glass
8067-05 8067-08 8067-11 8067-13 8067-30 8067-34 8067-38
89061-892EA 298.55 CAD
89061-892 89061-894 89061-898 89061-900 89061-908 89061-910 89061-912
Bearing, Trubore®, PTFE, 19 mm, Ace Glass Incorporated
Stirrers Stirring Tools
Self-aligning, lubricant-free PTFE Trubore® bearing for use with a 19mm polished glass stirring shaft
Complete item consists of PTFE inner Trubore® bearing, #25 Ace-Thred glass adapter, non-flaking PTFE compression saddle with o-ring and glass-reinforced PTFE bushing and lock nut with FETFE o-ring. The lock nut permits stirring in either direction without fear of unthreading. The compression saddle with o-ring maintains constant force with little attention. Vacuum down to 1mm of Hg. Not recommended for use with precision ground glass shafts.
Ordering information: For replacement "More Product Information