Photochemical Quartz Triple-Walled Immersion Well, Low Temperature, Ace Glass Incorporated
Supplier: Ace Glass
7858-07 7858-08 7858-13 7858-14 7858-42 7858-45 7858-81 7858-82 7858-84 7858-85 7858-88
89061-234EA 2904.41 CAD
89061-234 89061-236 89061-238 89061-240 89061-242 89061-244 89061-246 89061-248 89061-250 89061-252 89061-256
Photochemical Quartz Triple-Walled Immersion Well, Low Temperature, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Triple-walled, quartz, immersion well for use at temperatures as low as –78 °C
The innermost wall is held in place via a stopper and permits a carefully positioned, PTFE water inlet tube to extend below the lamp bottom. Inner tube is removable and may be interchanged with borosilicate glass tubes. One size tube fits both wells. Outer chamber consists of a 89061-250 drilled neoprene stopper, 89061-026 spacer O-Rings, metal water inlet, connecting tube and PTFE tubing.
With a standard taper 60/40 center inner joint. Outer two walls are permanently sealed together and the space betweenMore Product Information