O-Rings, Buna-N, Ace Glass Incorporated
Supplier: Ace Glass
7855-401 7855-402 7855-403 7855-404 7855-405 7855-406 7855-407 7855-408 7855-409 7855-410 7855-411 7855-412 7855-413 7855-414 7855-415 7855-416 7855-417 7855-418 7855-419 7855-420 7855-421 7855-422 7855-424 7855-426 7855-427 7855-428 7855-429 7855-430 7855-432 7855-434 7855-436 7855-438 7855-439 7855-440 7855-442 7855-444 7855-445 7855-446 7855-447 7855-448 7855-450 7855-470 7855-471 7855-472 7855-473 7855-474 7855-475 7855-476 7855-437 7855-441 7855-443 7855-451 7855-452 7855-453 7855-454 7855-455
89060-786PK 15.42 CAD
89060-786 89060-788 89060-790 89060-792 89060-794 89060-796 89060-798 89060-800 89060-802 89060-804 89060-806 89060-808 89060-810 89060-812 89060-814 89060-816 89060-818 89060-820 89060-822 89060-826 89060-828 89060-830 89060-832 89060-834 89060-836 89060-838 89060-840 89060-842 89060-844 89060-846 89060-848 89060-850 89060-852 89060-856 89060-858 89060-860 89060-862 89060-864 89060-866 89060-868 89060-872 89060-878 89060-880 89060-882 89060-884 89060-886 89060-888 89060-890 89215-112 89215-114 89215-116 89215-118 89215-120 89215-122 89215-124 89215-126
O-Rings, Buna-N, Ace Glass Incorporated
Seals Sealing Rings
Buna-N o-rings
Buna-N also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. It is generally resistant to fuels and oils, and is usually black or brown in color.
Order Now
SPECIFICATIONS
Environmentally Preferable