O-Rings, Silicone, Ace Glass Incorporated
Supplier: Ace Glass
7855-201 7855-202 7855-203 7855-204 7855-205 7855-206 7855-207 7855-208 7855-209 7855-210 7855-211 7855-212 7855-213 7855-214 7855-215 7855-216 7855-217 7855-218 7855-219 7855-220 7855-221 7855-222 7855-224 7855-226 7855-227 7855-228 7855-229 7855-230 7855-232 7855-234 7855-236 7855-238 7855-239 7855-240 7855-242 7855-244 7855-245 7855-246 7855-247 7855-248 7855-250 7855-264 7855-266 7855-270 7855-271 7855-272 7855-273 7855-274 7855-275 7855-276 7855-277 7855-278 7855-282 7855-283 7855-287 7855-289 7855-260 7855-288
89060-636PK 72.95 CAD
89060-636 89060-638 89060-640 89060-642 89060-644 89060-646 89060-648 89060-650 89060-652 89060-656 89060-658 89060-660 89060-662 89060-664 89060-666 89060-668 89060-670 89060-672 89060-674 89060-678 89060-680 89060-682 89060-684 89060-686 89060-688 89060-690 89060-692 89060-694 89060-696 89060-698 89060-700 89060-702 89060-704 89060-708 89060-710 89060-712 89060-714 89060-716 89060-718 89060-720 89060-722 89060-732 89060-734 89060-738 89060-740 89060-742 89060-744 89060-746 89060-748 89060-750 89060-752 89060-754 89060-758 89060-760 89060-762 89060-766 71000-160 71000-162 71000-164 71000-166
O-Rings, Silicone, Ace Glass Incorporated
Seals Sealing Rings
Silicone is a “rubber-like” compound of silicone, (itself a polymer) containing silicon with carbon, hydrogen, and oxygen. It is non-reactive and stable, with good temperature resistance. Generally red in color, Silicone O-Rings have an excellent compression set.