Ball and Socket Joints, Ace Glass Incorporated
Supplier: Ace Glass
7655-52 7655-04 7655-06 7655-10 7655-12 7655-16 7655-18 7655-22 7655-24 7655-28 7655-30 7655-34 7655-36 7655-40 7655-42 7655-46 7655-48 7655-60 7655-64 7655-70 7655-72 7655-76 7655-78 7655-54 7655-58 7655-66
89059-432EA 60.09 CAD
89059-432 89060-078 89060-080 89060-082 89060-084 89060-086 89060-088 89060-090 89060-092 89060-094 89060-096 89060-098 89060-100 89060-102 89060-104 89060-106 89060-108 89060-116 89060-118 89060-122 89060-124 89060-126 89060-128 89060-132 89060-134 89060-140
Ball and Socket Joints, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
Ball and socket joints allow flexibility in the angles of the pieces being joined.
  • Full-length
  • Standard wall
  • Precision ground
  • Fabricated from borosilicate glass
These full length, standard-wall spherical ball and socket joints are precision ground.
Order Now
SPECIFICATIONS
Environmentally Preferable