Inner Joints, with Ring, Rodaviss®, Ace Glass Incorporated
Supplier: Ace Glass
7613-03 7613-07 7613-11 7613-17
89059-706EA 49.98 CAD
89059-706 89059-708 89059-710 89059-712
Inner Joints, with Ring, Rodaviss®, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This is a standard taper inner ground glass joint with drip tip and ring on shank.
  • Forms a leak-tight seal
  • Drip tip
  • Ring on shank
These joints are designed for use with an outer member, O-Ring, and cap to form a leak-tight seal.
Order Now
SPECIFICATIONS
Environmentally Preferable