PTFE Sleeves for [ST] Joints, Ace Glass Incorporated
Supplier: Ace Glass
7551-02 7551-06 7551-08 7551-09 7551-10 7551-12 7551-14 7551-16 7551-18 7551-20 7551-22 7551-24
89059-388EA 57.4 CAD
89059-388 89059-390 89059-392 89059-394 89059-396 89059-398 89059-400 89059-402 89059-404 89059-406 89059-408 89059-410
PTFE Sleeves for [ST] Joints, Ace Glass Incorporated
NS Joint Sleeves and Connectors Joint Sleeves
Designed for use with [ST] inner member regular joints with only a slight mismatch, these PTFE sleeves are treated on the inner surface so that they can be cemented to glass with an epoxy adhesive
The sleeves may also be used uncemented, to replace lubricant at low pressure differentials. The sleeves are accurately shaped to 1:10 taper. They are rugged, with a wall thickness of approximately 0.38mm (0.015").
Order Now
SPECIFICATIONS
Environmentally Preferable