Air Sampling Manifolds, Ace Glass Incorporated
Supplier: Ace Glass
7488-15-48-10 7488-35-12-3 7488-35-48-6 7488-04-36 7488-04-48 7488-04-60 7488-04-72 7488-04-84 7488-05-36 7488-05-48 7488-05-60 7488-14-12-1 7488-14-12-2 7488-14-12-3 7488-14-12-4 7488-14-12-5 7488-14-12-6 7488-14-12-7 7488-14-12-8 7488-14-24-1 7488-14-24-2 7488-14-24-3 7488-14-24-4 7488-14-24-5 7488-14-24-6 7488-14-24-7 7488-14-36-1 7488-14-36-10 7488-14-36-2 7488-14-36-3 7488-14-36-4 7488-14-36-5 7488-14-36-6 7488-14-36-7 7488-14-36-8 7488-14-48-1 7488-14-48-10 7488-14-48-3 7488-14-48-4 7488-14-48-5 7488-14-48-6 7488-14-48-8 7488-14-60-4 7488-14-60-5 7488-14-60-6 7488-14-60-7 7488-14-72-5 7488-14-72-6 7488-14-72-7 7488-15-120-5 7488-15-120-6 7488-15-12-3 7488-15-12-5 7488-15-24-1 7488-15-24-3 7488-15-24-5 7488-15-24-6 7488-15-36-3 7488-15-36-5 7488-15-36-6 7488-15-48-2 7488-15-48-4 7488-15-48-8 7488-15-60-3 7488-15-60-6 7488-15-72-4 7488-15-72-5 7488-15-72-6 7488-15-84-6 7488-24-12 7488-24-120 7488-24-24 7488-24-36 7488-24-48 7488-24-72 7488-24-96 7488-25-12 7488-25-24 7488-25-48 7488-34-12-1 7488-34-12-2 7488-34-12-3 7488-34-12-4 7488-34-12-5 7488-34-12-6 7488-34-24-10 7488-34-24-2 7488-34-24-3 7488-34-24-4 7488-34-24-5 7488-34-24-6 7488-34-24-7 7488-34-24-8 7488-34-36-10 7488-34-36-2 7488-34-36-3 7488-34-36-4 7488-34-36-6 7488-34-36-8 7488-34-48-2 7488-34-48-3 7488-34-48-4 7488-34-48-5 7488-34-48-6 7488-34-48-8 7488-34-60-4 7488-34-60-5 7488-34-60-6 7488-34-72-4 7488-34-72-5 7488-34-72-6 7488-34-96-3 7488-34-96-8 7488-35-12-1 7488-35-12-5 7488-35-24-10 7488-35-24-2 7488-35-24-3 7488-35-24-4 7488-35-24-5 7488-35-24-6 7488-35-36-3 7488-35-36-4 7488-35-60-2 7488-35-60-6 7488-14-48-7 7488-14-24-8 7488-34-36-1 7488-35-24-11 7488-14-24-10 7488-35-24-1 7488-14-24-9 7488-14-60-12 7488-15-48-3
71001-552EA 2098.31 CAD
71001-552 71001-554 71001-556 89058-922 89058-924 89058-926 89058-928 89058-930 89058-936 89058-938 89058-940 89058-942 89058-944 89058-946 89058-948 89058-950 89058-952 89058-954 89058-956 89058-958 89058-960 89058-962 89058-964 89058-966 89058-968 89058-970 89058-972 89058-974 89058-976 89058-978 89058-980 89058-982 89058-984 89058-986 89058-988 89058-990 89058-992 89058-994 89058-996 89058-998 89059-000 89059-002 89059-004 89059-006 89059-008 89059-010 89059-012 89059-014 89059-016 89059-018 89059-020 89059-022 89059-024 89059-026 89059-028 89059-030 89059-032 89059-034 89059-036 89059-038 89059-040 89059-042 89059-044 89059-046 89059-048 89059-050 89059-052 89059-054 89059-056 89059-058 89059-060 89059-062 89059-064 89059-066 89059-068 89059-070 89059-072 89059-074 89059-076 89059-078 89059-080 89059-082 89059-084 89059-086 89059-088 89059-090 89059-092 89059-094 89059-096 89059-098 89059-100 89059-102 89059-104 89059-106 89059-108 89059-110 89059-112 89059-114 89059-116 89059-118 89059-120 89059-122 89059-124 89059-126 89059-128 89059-130 89059-132 89059-134 89059-136 89059-138 89059-140 89059-142 89059-144 89059-146 89059-148 89059-150 89059-152 89059-154 89059-156 89059-158 89059-160 89059-162 89059-164 89059-166 89059-168 89084-232 89084-236 89084-240 89084-280 89097-164 10767-304 10767-936 10767-938 10767-940
Air Sampling Manifolds, Ace Glass Incorporated
Manifolds
These straight air sampling manifolds are available in a variety of different configurations and various lengths
An appropriate size threaded bushing or coupling can used on the threaded end(s), as well as on the tube ends. Models with sampling port(s) are supplied with the appropriate size bushing. All manifolds are fabricated from borosilicate glass.
Ordering information: If connecting thread to thread, users cannot be certain the ports will be positioned in the desired direction. Therefore, special configurations are available via special order. Contact a Avantor representative for details.