Extraction Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
6846-08 6846-11 6846-14 6846-15 6846-24 6846-25 6846-30 6846-50
89056-090EA 1112.4 CAD
89056-090 89056-092 89056-094 89056-096 89056-098 89056-100 89056-102 89056-104
Extraction Apparatus, Ace Glass Incorporated
Extractors Liquid-Liquid Extractors
A liquid-liquid extractor for use with lighter- or heavier-than-water extracting solvents
When working with solvents lighter than water, the glass dispersion tube adapter is inserted into the extractor body and the #25 Ace-Thred on the body and #7 Ace-Thred near the bottom are plugged. Solvent flask is heated and vapors rise to condenser through glass elbow tube connected by two #25 threads. Condensed solvent fills adapter tube stem and is dispersed upward through sample from capillary on bottom of stem, thereby increasing solvent surface-area-to-volume ratio. Solvent overflows thru PTFEMore Product Information