Extraction Thimbles, Glass, Ace Glass
Supplier: Ace Glass
6813-02 6813-04 6813-06 6813-08 6813-10 6813-12 6813-14 6813-16 6813-18 6813-20 6813-22 6813-24 6813-26 6813-28 6813-30
89056-030EA 171.46 CAD
89056-030 89056-032 89056-034 89056-036 89056-038 89056-040 89056-042 89056-044 89056-046 89056-048 89056-050 89056-052 89056-054 89056-056 89056-058
Extraction Thimbles, Glass, Ace Glass
Extractors Extraction Thimbles
These thimbles come with an ACE fritted disc sealed in
Can be used in any standard Soxhlet extraction apparatus. The sizes below correspond to the size specifications of extraction bodies, and will fit those units. Available in five different porosities. Fabricated from borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable