Soxhlet and Soxhlet-Type Extractors, Ace Glass
Supplier: Ace Glass
6776-02 6776-04 6776-06 6776-10
89055-986EA 252.32 CAD
89055-986 89055-988 89055-990 89055-992
Soxhlet and Soxhlet-Type Extractors, Ace Glass
Extractors Soxhlet and Soxhlet-Type Extractors
This Micro Soxhlet Extractor is designed for small chemical applications
Improved design permits greater condensing capacity. The condenser jacket length is approximately 139mm. The joint between the condenser and the extractor is a [ST] 24/25. Flask joint is [ST] 14/20. Designed to accommodate paper thimble, size 10x50mm. The flask is 30mL capacity. Hose connections are for use with 5/16 or 3/8" I.D. tubing. Fabricated from borosilicate glass.
Ordering information: Complete item includes 89055-986 condenser, 89055-988 extractor, and 89055-990 flask. Components may also be purchased separately.