Extraction Apparatus, Soxhlet with Friedrichs Condenser, Ace Glass
Supplier: Ace Glass
6718-15 6718-23 6718-29
89055-956EA 1372.72 CAD
89055-956 89055-958 89055-960
Extraction Apparatus, Soxhlet with Friedrichs Condenser, Ace Glass
Extractors Soxhlet and Soxhlet-Type Extractors
The improved design of this Soxhlet extractor with Friedrichs condenser permits greater condensing capacity
Cycling rates may be doubled over conventional style extractors. Fabricated from borosilicate glass.
Ordering information: This apparatus comes complete with condenser, extractor, and flask listed below. These components may also be purchased separately.
Order Now
SPECIFICATIONS
Environmentally Preferable