Freeze Drying Vessels with Screw Cap, Ace Glass
Supplier: Ace Glass
6696-20 6696-22 6696-24
89055-770EA 173.41 CAD
89055-770 89055-778 89055-786
Freeze Drying Vessels with Screw Cap, Ace Glass
Sample Containers General Purpose Containers
This freeze drying vessel has a GCMI-24-410 bottle thread at top and comes complete with a PTFE lined cap.
The neck also fits inside a #25 Ace-Thred bushing, so it can be used with a 89055-766 umbrella manifold. After drying the material, the vessel can be capped and stored.
Ordering information: For bushings, see part 89059-226. Working capacity of flasks is about one-half the volume listed.
Order Now
SPECIFICATIONS
Environmentally Preferable