Distillation Apparatus, Cyanide, Ace Glass
Supplier: Ace Glass
6556-05 6556-07 6556-10 6556-12 6556-13 6556-23 6556-26 6556-27 6556-33
89055-594EA 1854.62 CAD
89055-594 89055-596 89055-598 89055-600 89055-602 89055-604 89055-606 89055-608 89055-610
Distillation Apparatus, Cyanide, Ace Glass
Distillation Components Distillation Sets
This cyanide distillation apparatus is used in testing for soluble and insoluble cyanides in water
In Model A, two #11 Ace-Thred bushings, back to back, hold a threaded glass adapter (which has side outlet tube with hose barb) to the still head by compressing PTFE ferrules against the glass dispersion tube. The adapter then joins to the trap via [ST] 19/22 joints. Because of this feature, the dispersion tube depth is adjustable inside the trap, and the fritted end need not be in contact with caustic solutions. This will extend the life of the frit and assure consistent dispersion for aMore Product Information