Jacketed Cylindrical Reaction Flask with Duran Flange, Ace Glass Incorporated
Supplier: Ace Glass
6523-12 6523-14 6523-16 6523-19 6523-22 6523-27 6523-32 6523-37 6524-13 6524-15 6524-17 6524-20 6523-30 6523-312 6523-314 6523-316 6523-319 6524-313 6524-315 6524-317 6524-320
89055-206EA 2011.44 CAD
89055-206 89055-208 89055-210 89055-212 89055-214 89055-216 89055-218 89055-220 89055-222 89055-224 89055-226 89055-228 89083-550 10767-176 10767-178 10767-180 10767-290 10767-292 10767-294 10767-296 10767-298
Jacketed Cylindrical Reaction Flask with Duran Flange, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
The rugged cylindrical reaction flask with Duran style flat o-ring groove flange, is for use with quick release clamp
Fabricated from heavy wall borosilicate glass. Inlet and outlet connections on jacket, sealed tangentially, are o-ring ball joints. Small reactors 1 to 6 L use 28/15 spherical joints and 10 L and larger us 35/25 o-ring joints. Ten liter and larger are supplied with capacity reference graduations. Each flask is supplied with one CAPFE, PTFE encapsulated silicone, o-ring. Flasks are available with a variety of valve types. Maximum Jacket Pressure: 5 PSI (.34 Bar). JacketMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable