Unjacketed Filter Reactor, Pressure Rated to 35 psi, Ace Glass Incorporated
Supplier: Ace Glass
6384-05 6384-11 6384-115 6384-121 6384-125 6384-129 6384-133 6384-137 6384-15 6384-19 6384-225 6384-23 6384-231 6384-235 6384-239 6384-243 6384-247 6384-27
89054-088EA 1013.13 CAD
89054-088 89054-090 89054-092 89054-094 89054-096 89054-098 89054-100 89054-102 89054-104 89054-106 89054-108 89054-110 89054-112 89054-114 89054-116 89054-118 89054-120 89054-122
Unjacketed Filter Reactor, Pressure Rated to 35 psi, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
These heavy wall glass reactors are available with a 60mm (2.4"), 100mm (4"), or 150mm (6") diameter Duran style flange with O-Ring groove at top
Ordering Information: Complete item consists of reactor body with 60mm CAPFE O-Ring, head with Ace-Threds, clamp, PTFE bottom threaded adapter and valve with CAPFE O-Ring, retainer ring, 350µm polypropylene screen support, 100µm polyethylene support, adapter, bearing, stir rod, paddle agitator, condenser, addition funnel, and #7 Ace-Thred bushing. Size 80 bottom adapter also comes with a perforated glass plate support.
Connects to a head withMore Product Information