Flash Chromatography Column, [SJ] Joints, Ace Glass Incorporated
Supplier: Ace Glass
5872-02 5872-03 5872-04 5872-05 5872-06 5872-07 5872-08 5872-09 5872-10 5872-12 5872-13 5872-14 5872-15 5872-16 5872-17 5872-18 5872-19 5872-20 5872-22 5872-23 5872-24 5872-25 5872-26 5872-27 5872-28 5872-29 5872-30 5872-31 5872-32 5872-33 5872-34 5872-35 5872-36 5872-37 5872-41 5872-42 5872-43 5872-44 5872-45 5872-46 5872-47 5872-48 5872-49 5872-50 5872-51 5872-52 5872-54 5872-53 5872-55 5872-56 5872-57 5872-58 5872-59 5872-60 5872-61 5872-62 5872-63 5872-64 5872-65 5872-66 5872-67 5872-68 5872-69 5872-70 5872-71 5872-72 5872-73 5872-74 5872-75 5872-76 5872-80 5872-81 5872-82 5872-86 5872-87 5872-88 5872-90 5872-97 5872-98
89053-262EA 382.67 CAD
89053-262 89053-264 89053-266 89053-268 89053-270 89053-272 89053-274 89053-276 89053-278 89053-280 89053-282 89053-284 89053-286 89053-288 89053-290 89053-292 89053-294 89053-296 89053-298 89053-300 89053-302 89053-304 89053-306 89053-308 89053-310 89053-312 89053-314 89053-316 89053-318 89053-320 89053-322 89053-324 89053-326 89053-328 89053-330 89053-332 89053-334 89053-336 89053-338 89053-340 89053-342 89053-344 89053-346 89053-348 89053-350 89053-352 89053-354 89053-356 89053-358 89053-360 89053-362 89053-364 89053-366 89053-368 89053-370 89053-372 89053-374 89053-376 89053-378 89053-380 89053-382 89053-384 89053-386 89053-388 89053-390 89053-392 89053-394 89053-396 89053-398 89053-400 89053-402 89053-404 89053-406 89053-408 89053-410 89053-412 89053-414 89053-416 89053-418
Flash Chromatography Column, [SJ] Joints, Ace Glass Incorporated
Chromatography Columns
This is a simple absorption chromatography column for rapid preparative separations as described by W.C
Column and reservoir (only) are epoxy coated for safety. Flow control adapter is uncoated. All are fabricated from borosilicate glass.
Still, M. Kahn and A. Mitra of Columbia University Department of Chemistry. Allows separations of samples from 0.01 to 10.0 g³ in 10 to 15 minutes. Reservoir connects between column and flow control adapter. Joints on column, reservoir and adapter are [SJ]35/20, [SJ] 50/30, or [SJ] 75/50. Columns available with or without Porosity B (70 to 100 µm)More Product Information