Buret, Weighing, Ace Glass Incorporated
Supplier: Ace Glass
5625-05 5625-07 5625-08 5625-06
89051-900EA 139.43 CAD
89051-900 89051-904 89051-906 89051-916
Buret, Weighing, Ace Glass Incorporated
Burettes
This Smith type weighing buret is used to dispense precise quantities of liquids
Capacity is 10 mL. The wide base adds stability when the buret is placed on a balance pan. Complete apparatus consists of a flask, buret, and cap stopper. Has a standard taper 19/10 cap-type stopper joint, and a standard taper 12/10 joint at the bottom. The overall height has been reduced to a minimum so that it can be used on single pan balances.
Ordering information: You can purchase, flask, buret, and cap stopper separately, or a complete item that includes all three.