Bottle, Storage, Heavy Wall, Ace Glass
Supplier: Ace Glass
5555-02 5555-03 5555-06 5555-22 5555-23 5555-24 5555-25 5555-32 5555-33
89051-868EA 153.73 CAD
89051-868 89051-870 89051-872 89051-874 89051-876 89051-878 89051-880 89051-882 89051-884
Bottle, Storage, Heavy Wall, Ace Glass
Bottles Glass Bottles
This heavy wall glass storage bottle with #15 Ace-Thred neck is intended only for storage of low-boiling liquids from –20 °C to ambient.
The plastic coating on this bottle is largely UV absorbent (up to 380 nm). May be autoclaved in steam up to a maximum of 135 °C. Not suitable for pressure, vacuum, freezing, or for microwave operation. Do not use with distilled water or vacuum for cooling.
Designed to resist internal pressures up to 100 psig in the stated temperature range. The low pressure rating is based primarily on the rating for PTFE/T.F.E. When purged with an inert gas beforeMore Product Information