Drying Tube Adapters, Ace Glass Incorporated
Supplier: Ace Glass
5170-05 5170-10 5170-15 5170-40
89051-350EA 158.95 CAD
89051-350 89051-352 89051-354 89051-356
Drying Tube Adapters, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
These U-shaped drying tube adapters are manufactured from borosilicate glass.
  • U-shaped
  • Inner joint on one end
  • One flared open end
Adapters feature an inner joint on one end and flared open end for a No. 3 rubber stopper and 6.4mm diameter x 50.8mm length (¼ x 2") glass tube. The stopper/tube can be purchased separately.